Learn More
EVOSEP EVOSEP Valve Stator, Single, 0.1 kg
Supplier: EVOSEP EV1010
The EVOSEP Valve 6 stator is a single stator designed for use with EV1009 systems.
- Single stator
- Weighs 0.1 kg
- Classified as a wear part