Learn More
Poltex Evotech Scope Connector Drying Rack
SDP
Catalog No. 22000642
Change view
Click to view available options
Closure Included:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Included
22-000-642 Yes
22-000-641 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-642 Supplier Poltex Supplier No. SCOPCONBX
Add to Cart
Add to Cart
This item is not returnable. View return policy
Evotech Scope Connector Drying Rack - Works with Evotech or similar scope connectors. Super durable constructions eliminates sag of long drying slots.
  • Works with Evotech or similar scope connectors.
  • Choose from two versions: with wire box storage space or without.
  • See image for dimensions.
  • Super durable 1/2 in. and 1/4 in. thick acrylic constructions eliminates sag of long drying slots.
Specifications
Closure Included Yes
Material Acrylic
No. of Racks 14
Product Type Drying Rack