Learn More
Andwin Scientific EVT300 Mini-Gel Power Supply. Fixed voltage dual power supply. Voltage: 70 or 125 volts. Maximum current: 250 milliamps.
SDP
Catalog No. 50820187
Encompass_Preferred
EVT300 Mini-Gel Power Supply. Fixed voltage dual power supply. Voltage: 70 or 125 volts. Maximum current: 250 milliamps.
Catalog No. 50-820-187 Supplier Andwin Scientific Supplier No. 34065090
EVT300 Mini-Gel Power Supply. Fixed voltage dual power supply. Voltage: 70 or 125 volts. Maximum current: 250 milliamps.