Learn More
Selleck Chemical LLC Vactosertib 5mg 1352608-82-2 TEW-7197
SDP
Supplier:  Selleck Chemical LLC S75305MG
Encompass_Preferred
Vactosertib ¿(TEW-7197, EW-7197) is a highly potent, selective, and orally bioavailable TGF-? receptor ALK4/ALK5 inhibitor with IC50 of 13 nM and 11 nM, respectively. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1107271