Learn More
Aviva Systems Biology Corp EWSR1 Antibody - middle region (ARP38594_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38594_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EWSR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EWSR1. Alternate names: EWS, bK984G1.4ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56719