Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ EWSR1 Split FISH Probe

Catalog No. 89024951 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-951 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-951 Supplier Abnova™ Supplier No. FS0003
Add to Cart
Add to Cart

Includes

DAPI Counterstain (1500ng/mL) 250μL

Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Content And Storage Store at 4°C in the dark.
Label or Dye FITC, Texas Red
For Use With (Application) Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Location 22q12, 22q12
Probe EWSR1, EWSR1
Product Type EWSR1 Split FISH Probe
Quantity 200 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.