Learn More
Apexbio Technology LLC EX 527 (SEN0014196) 49843-98-3 25mg
SDP
Catalog No. 5000061548
Encompass_Preferred
EX 527 (SEN0014196 CAS 49843-98-3) is a small-molecule inhibitor targeting the catalytic activity of the SIRT1 enzyme By selectively inhibiting SIRT1 EX 527 increases acetylation of the tumor suppressor protein p53 following DNA damage Research using etoposide-treated cells indicates that inhibition by EX 527 does not significantly alter cellular viability growth or p53-regulated gene transcription EX 527 demonstrates high selectivity for SIRT1 showing approximately 200 500-fold greater specificity compared to homologous enzymes SIR2 and SIRT3 Additionally EX 527 exists as a racemic mixture separable into distinct stereoisomers (EX-242 EX-243) via chiral HPLC aiding structure-activity studies
Catalog No. 50-000-61548 Supplier Apexbio Technology LLC Supplier No. A418125
EX 527 (SEN0014196 CAS 49843-98-3) is a small-molecule inhibitor targeting the catalytic activity of the SIRT1 enzyme By selectively inhibiting SIRT1 EX 527 increases acetylation of the tumor suppressor protein p53 following DNA damage Research using etoposide-treated cells indicates that inhibition by EX 527 does not significantly alter cellular viability growth or p53-regulated gene transcription EX 527 demonstrates high selectivity for SIRT1 showing approximately 200 500-fold greater specificity compared to homologous enzymes SIR2 and SIRT3 Additionally EX 527 exists as a racemic mixture separable into distinct stereoisomers (EX-242 EX-243) via chiral HPLC aiding structure-activity studies