Learn More
Abcam E x -527, SIRT1 Inhibitor, 10MG
Catalog No. 502706554
Encompass_Preferred
MW 248.71 Da, Purity >97%. Selective, cell-permeable SIRT1 Inhibitor. (IC₅₀= 98 nM, 19.6, 48.7 and > 100 μM for SIRT1, SIRT2, SIRT3, HDAC and NADase respectively). Shows no effect at other HDACs or SIRTs. Enhances p53 acetylation in response to DNA damaging agents.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6554 Supplier Abcam Supplier No. AB14150610MG
MW 248.71 Da, Purity >97%. Selective, cell-permeable SIRT1 Inhibitor. (IC₅₀= 98 nM, 19.6, 48.7 and > 100 μM for SIRT1, SIRT2, SIRT3, HDAC and NADase respectively). Shows no effect at other HDACs or SIRTs. Enhances p53 acetylation in response to DNA damaging agents.
The product is subject to the following: Abcam Restricted Use Statement