Learn More
TARGETMOL CHEMICALS INC EX-A5758 5MG
SDP
Catalog No. 502578922
Encompass_Preferred
Also available in 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EX-A5758 is a novel putative small molecule nNOS-NOS1AP inhibitor suppressing inflammatory nociception and chemotherapy-induced neuropathic pain and synergizes with paclitaxel to reduce tumor cell viability. purity: 98%
Catalog No. 50-257-8922 Supplier TARGETMOL CHEMICALS INC Supplier No. T99555MG
Also available in 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EX-A5758 is a novel putative small molecule nNOS-NOS1AP inhibitor suppressing inflammatory nociception and chemotherapy-induced neuropathic pain and synergizes with paclitaxel to reduce tumor cell viability. purity: 98%