Learn More
Sigma Aldrich Fine Chemicals Biosciences EX-CELL 420 Serum-Free Medium for Insect Cells, 1000 mL
Catalog No. NC1898243
Encompass
EX-CELL 420 is a complete medium developed and optimized for the serum-free growth of Sf9 and Sf21 insect cell lines. Cells can be subcultured directly into EX-CELL 420 from serum-free or seum-supplemented media without adaptation. Cultures in EX-CELL 420 routinely reach cell densities greater than 1 × 107 cells/mL with greater than 95% viability. Suspension cultures can be maintained, without refeeding, for more than 10 days. Sf9 and Sf21 cells have been carried for more than 20 passages in EX-CELL 420 with no loss of viability. Protein expression and virus production are improved over serum-containing media.
Catalog No. NC1898243 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14420C1000ML
EX-CELL 420 is a complete medium developed and optimized for the serum-free growth of Sf9 and Sf21 insect cell lines. Cells can be subcultured directly into EX-CELL 420 from serum-free or seum-supplemented media without adaptation. Cultures in EX-CELL 420 routinely reach cell densities greater than 1 × 107 cells/mL with greater than 95% viability. Suspension cultures can be maintained, without refeeding, for more than 10 days. Sf9 and Sf21 cells have been carried for more than 20 passages in EX-CELL 420 with no loss of viability. Protein expression and virus production are improved over serum-containing media.