Learn More
Sigma Aldrich Fine Chemicals Biosciences EX-CELL Antifoam
Catalog No. NC1497386
Encompass
EX-CELL ANTIFOAM is a USP Grade non-ionic emulsion (simethicone) designed for use in the pharmaceutical and veterinary biological industries to aid in the control of foaming typically associated with the use of culture medium in bioreactors.
Catalog No. NC1497386 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 59920C5B
EX-CELL ANTIFOAM is a USP Grade non-ionic emulsion (simethicone) designed for use in the pharmaceutical and veterinary biological industries to aid in the control of foaming typically associated with the use of culture medium in bioreactors.