Learn More
Sigma Aldrich Fine Chemicals Biosciences EX-CELL(R) CD CHO Serum-Free Medium for CHO Cells, Chemically Defined Animal-component free, without L-glutamine, without hypoxanthine, without thymidine, liquid, sterile-filtered, suitable for cell culture | MFCD01634638 | 1000ML
Catalog No. NC0514251
Encompass
EX-CELL(R) CD CHO Serum-Free Medium for CHO Cells, Chemically Defined Animal-component free, without L-glutamine, without hypoxanthine, without thymidine, liquid, sterile-filtered, suitable for cell culture | MFCD01634638 | 1000ML
Catalog No. NC0514251 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14360C1000ML
EX-CELL(R) CD CHO Serum-Free Medium for CHO Cells, Chemically Defined Animal-component free, without L-glutamine, without hypoxanthine, without thymidine, liquid, sterile-filtered, suitable for cell culture | MFCD01634638 | 1000ML