Learn More
Chemglass Life Sciences Clamp, Column, Extra Large, 140mm to 165mm Clamping Range
SDP
Catalog No. CG922104
Encompass_Preferred
Clamp, Column, Extra Large, 140mm to 165mm Clamping Range. Ideal for Chromatography columns and large cylindrical glassware, the column clamp is constructed of stainless steel. Large, flat thumb screw on the worm drive permits tightening and removing.
Catalog No. CG922104 Supplier Chemglass Life Sciences Supplier No. CG922104
Clamp, Column, Extra Large, 140mm to 165mm Clamping Range. Ideal for Chromatography columns and large cylindrical glassware, the column clamp is constructed of stainless steel. Large, flat thumb screw on the worm drive permits tightening and removing.