Learn More
Takara Bio EX TAQ (MG2+ FREE BUFFER) 250
SDP
Catalog No. 50443988
Encompass
PCR Reagents; Takara; Ex Taq DNA Polymerase, dNTPs, magnesium-free buffer, and separate tube of MgCl2 included; 250U, Excellent for amplifying fragments from problem organisms
Catalog No. 50-443-988 Supplier Takara Bio Supplier No. RR01AM
PCR Reagents; Takara; Ex Taq DNA Polymerase, dNTPs, magnesium-free buffer, and separate tube of MgCl2 included; 250U, Excellent for amplifying fragments from problem organisms