Learn More
Takara Bio EX TAQ POLYM HOT-START 250U
SDP
Catalog No. 50443979
Encompass
PCR Reagents; Takara; Hot-Start Ex Taq DNA Polymerase; 250U; reduces background, increased specificity, permits room-temperature reaction assembly
Catalog No. 50-443-979 Supplier Takara Bio Supplier No. RR006A
PCR Reagents; Takara; Hot-Start Ex Taq DNA Polymerase; 250U; reduces background, increased specificity, permits room-temperature reaction assembly