Learn More
Medchemexpress LLC HY-112769 10mg Medchemexpress, EX229 CAS:1219739-36-2 Purity:>98%
SDP
Catalog No. 501969165
Encompass_Preferred
Medchemexpress, HY-112769 10mg EX229 CAS:1219739-36-2 EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9165 Supplier Medchemexpress LLC Supplier No. HY11276910MG
Medchemexpress, HY-112769 10mg EX229 CAS:1219739-36-2 EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.