Learn More
Selleck Chemical LLC ex229 (compound 991) 5mg 1219739-36-2
SDP
Catalog No. 501948263
Encompass_Preferred
EX229 (compound 991) is a potent AMPK activator that is 5-10-fold more potent than A769662 in activating AMPK. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8263 Supplier Selleck Chemical LLC Supplier No. S86545MG
Add to Cart
Add to Cart
EX229 (compound 991) is a potent AMPK activator that is 5-10-fold more potent than A769662 in activating AMPK. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.