Learn More
Selleck Chemical LLC Selisistat (EX 527) 10mM/1mL 49843-98-3 SEN0014196
SDP
Supplier:  Selleck Chemical LLC S154110MM/1ML
Encompass_Preferred
Selisistat (EX 527, SEN0014196) is a potent and selective SIRT1 inhibitor with IC50 of 38 nM in a cell-free assay, exhibits >200-fold selectivity against SIRT2 and SIRT3. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1278542