Learn More
Electron Microscopy Sciences Exact Volume Pipet 60 µl, 2.5 mm, 6.5 cm Stem diameter
SDP
Catalog No. 501899421
Encompass_Preferred
These pipets offer the exact volume of liquid just one squeeze to the bulb. Quick and economical for disposable pipets, without the need of expensive pipetting devices. These pipets come non-sterile.
Volume: 2.5 mm
Stem diameter: 6.5 cm
OAL: Bulk
Catalog No. 50-189-9421 Supplier Electron Microscopy Sciences Supplier No. 7096932
These pipets offer the exact volume of liquid just one squeeze to the bulb. Quick and economical for disposable pipets, without the need of expensive pipetting devices. These pipets come non-sterile.
Volume: 2.5 mm
Stem diameter: 6.5 cm
OAL: Bulk