Learn More
Hanna Pharmaceutical Supply Co., Inc. EXACTAMX EMPTY BAG 1000ML
Catalog No. NC1036789
Encompass_Preferred
Due to recent natural disasters, production of sterile water has been negatively impacted.  Please expect long lead times on these itemsEVA (ethylene vinyl acetate) Used for large-volume ster.solutions. Non-reopening clamp attached. For use with EXACTAMIX Compounder. Sterile Fluid Path. Non-DEHP. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC1036789 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BAXH938739
Due to recent natural disasters, production of sterile water has been negatively impacted.  Please expect long lead times on these itemsEVA (ethylene vinyl acetate) Used for large-volume ster.solutions. Non-reopening clamp attached. For use with EXACTAMIX Compounder. Sterile Fluid Path. Non-DEHP. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.