Learn More
Genevac Inc Genevac EXALT Software/Starter Kit, Controlled Evaporative Crystallization, Polymorph Screening, 1 Kit
Catalog No. NC3656467
Encompass
The GENEVAC LTD EXALT SOFTWARE/STARTER KIT is a specialized toolkit designed for controlled evaporative crystallization studies, offering scientists efficient polymorph screening and non-destructive crystal production from minimal sample sizes. It integrates seamlessly with Genevac HT series and EZ-2 4.0 evaporators, providing precise evaporation control and customizable parameters, making it ideal for late-stage medicinal chemistry applications and solvent screening.
Catalog No. NC3656467 Supplier Genevac Inc Supplier No. 105100
Add to Cart
Add to Cart
Request a Quote
The GENEVAC LTD EXALT SOFTWARE/STARTER KIT is a specialized toolkit designed for controlled evaporative crystallization studies, offering scientists efficient polymorph screening and non-destructive crystal production from minimal sample sizes. It integrates seamlessly with Genevac HT series and EZ-2 4.0 evaporators, providing precise evaporation control and customizable parameters, making it ideal for late-stage medicinal chemistry applications and solvent screening.