Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ EXAMET-5 Reflected and Transmitted Illumination
SDP

Catalog No. 01189258 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Eyepiece:
WF 10X/23 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Eyepiece
01-189-258 WF 10X/23 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-258 Supplier ACCU-SCOPE™ Supplier No. 14351
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The UNITRON™ Versamet 4 features a trinocular viewing head. Eyepiece accepts a 24 mm reticle infinity M LWD plan achromat 5x,10x, 20x, and semi-apo 50x objectives, encoded quintuple nosepiece with DIC slot, ECO function, 12 volt 100 watt halogen illuminator, coaxial coarse and fine focusing controls.

  • Trinocular viewing head, inclined 30°
  • Viewing split 100% binocular, or 50% binocular / 50% trinocular
  • Widefield 10x eyepieces with a 23 mm field of view
  • Infinity LWD M plan achromat 5x, 10x, 20x objectives, Infinity LWD M Plan semi-Apo 50x objective
  • Reversed quintuple nosepiece
  • Interpupillary distance adjustment 54 mm to 75 mm
  • Mechanical Stage (175 x 145 mm), 76 mm (X) x 42 mm (Y) movement range
  • Slide holder not included
  • Low position coaxial focusing system with tension adjustment
  • Observation height of up to 30 mm
  • 5-watt variable LED reflected illuminator with field and aperture diaphragm
  • Condenser with swing-out top lens, condenser iris diaphragm, N.A. 0.9
  • Universal power supply (90 to 240 v)

Specifications

Eyepiece WF 10X/23 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.