Learn More
Medchemexpress LLC Exatecan | 171335-80-1 | 99.8% | 435.45 | 250 MG
SDP
Catalog No. 5000350144
Encompass_Preferred
Exatecan is a DNA topoisomerase I inhibitor with an IC50 of 2.2 μM (0.975 μg/mL). It can be used in cancer research.
  • Acts as a DNA topoisomerase I inhibitor
  • Effective at an IC50 of 2.2 μM (0.975 μg/mL)
  • Suitable for cancer research
Catalog No. 50-003-50144 Supplier Medchemexpress LLC Supplier No. 250MGHY13631
Exatecan is a DNA topoisomerase I inhibitor with an IC50 of 2.2 μM (0.975 μg/mL). It can be used in cancer research.
  • Acts as a DNA topoisomerase I inhibitor
  • Effective at an IC50 of 2.2 μM (0.975 μg/mL)
  • Suitable for cancer research