Learn More
Medchemexpress LLC Exatecan (Synonyms: DX-8951) 5mg, Solid, | CAS No. : 171335-80-1
SDP
Catalog No. NC3666762
Encompass_Preferred
Exatecan (DX-8951) is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL), and can be used in cancer research.
Catalog No. NC3666762 Supplier Medchemexpress LLC Supplier No. HY136315MG
Exatecan (DX-8951) is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL), and can be used in cancer research.