Learn More
Medchemexpress LLC Exatecan-amide-cyclopropanol | 2414254-34-3 | C28H26FN3O6 | 25 MG

Supplier: Medchemexpress LLC 25MGHY150401
Exatecan-amide-cyclopropanol is an anticancer agent that demonstrates anti-proliferation activity against SK-BR-3 and U87 cells. This product is for research use only and not intended for patient use.
- Exhibits anti-proliferation activity against SK-BR-3 cells with an IC50 of 0.12 nM.
- Exhibits anti-proliferation activity against U87 cells with an IC50 of 0.23 nM.
- Targeted as an ADC Cytotoxin and Topoisomerase inhibitor.
- Involves Antibody-drug Conjugate/ADC Related and Cell Cycle/DNA Damage pathways.
- Soluble in DMSO: 2.73 mg/mL (5.25 mM) with ultrasonic and warming and heating to 80°C.
- Molecular weight: 519.52.
- Belongs to the Camptothecins class.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.