Learn More
Medchemexpress LLC Exatecan-amide-cyclopropanol | 2414254-34-3 | C28H26FN3O6 | 5 MG
SDP
Catalog No. 5000375679
Encompass_Preferred
Exatecan-amide-cyclopropanol is an anticancer agent that demonstrates anti-proliferation activity against SK-BR-3 and U87 cells, with IC50 values of 0.12 and 0.23 nM, respectively. This product is intended for research use only.
  • Acts as an anticancer agent
  • Exhibits anti-proliferation activity against SK-BR-3 and U87 cells
  • Has IC50 values of 0.12 and 0.23 nM
  • Targets ADC Cytotoxin and Topoisomerase pathways
  • Related to antibody-drug conjugate/ADC and cell cycle/DNA damage
Catalog No. 50-003-75679 Supplier Medchemexpress LLC Supplier No. HY1504015MG
Exatecan-amide-cyclopropanol is an anticancer agent that demonstrates anti-proliferation activity against SK-BR-3 and U87 cells, with IC50 values of 0.12 and 0.23 nM, respectively. This product is intended for research use only.
  • Acts as an anticancer agent
  • Exhibits anti-proliferation activity against SK-BR-3 and U87 cells
  • Has IC50 values of 0.12 and 0.23 nM
  • Targets ADC Cytotoxin and Topoisomerase pathways
  • Related to antibody-drug conjugate/ADC and cell cycle/DNA damage