Learn More
Medchemexpress LLC HY-13631AS 10mg , Exatecan-d5 (mesylate) CAS: Purity:>98%
SDP
Catalog No. 502261407
Encompass_Preferred
HY-13631AS 10mg Exatecan-d5 (mesylate) CAS: Exatecan-d5 mesylate (DX8951f-d5) is deuterium labeled Exatecan mesylate. Exatecan mesylate is a DNA topoisomerase I inhibitor, with an IC50 of 0.975 μg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1407 Supplier Medchemexpress LLC Supplier No. HY13631AS10MG
HY-13631AS 10mg Exatecan-d5 (mesylate) CAS: Exatecan-d5 mesylate (DX8951f-d5) is deuterium labeled Exatecan mesylate. Exatecan mesylate is a DNA topoisomerase I inhibitor, with an IC50 of 0.975 μg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.