Learn More
Medchemexpress LLC Exatecan-d5 mesylate | 2819276-88-3 | 99.8% | 536.58 | 1 MG
SDP
Catalog No. 5000352240
Encompass_Preferred
Exatecan-d5 mesylate is a deuterium-labeled version of Exatecan mesylate. It is a DNA topoisomerase I inhibitor with an IC50 of 2.2 μM (0.975 μg/mL) and is used in cancer research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules, serving as tracers for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-52240 Supplier Medchemexpress LLC Supplier No. HY13631AS1MG
Exatecan-d5 mesylate is a deuterium-labeled version of Exatecan mesylate. It is a DNA topoisomerase I inhibitor with an IC50 of 2.2 μM (0.975 μg/mL) and is used in cancer research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules, serving as tracers for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.