Learn More
Medchemexpress LLC Exatecan-d5 mesylate | 2819276-88-3 | 99.8% | 536.58 | 25 MG
SDP
Catalog No. 5000348032
Encompass_Preferred
Exatecan-d5 (DX8951f-d5) mesylate is deuterium labeled Exatecan mesylate (HY-13631A). Exatecan mesylate is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). It can be used in cancer research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-48032 Supplier Medchemexpress LLC Supplier No. 25MGHY13631AS
Exatecan-d5 (DX8951f-d5) mesylate is deuterium labeled Exatecan mesylate (HY-13631A). Exatecan mesylate is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). It can be used in cancer research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.