Learn More
Medchemexpress LLC Exatecan-d5 mesylate (DX8951f-d5) | 2819276-88-3 | 99.8% | 536.58 | 100 MG
SDP
Catalog No. 5000352348
Encompass_Preferred
Exatecan-d5 mesylate (DX8951f-d5) is a deuterium-labeled compound used as a tracer and internal standard for quantitative analysis by methods like NMR, GC-MS, or LC-MS. Its unlabeled form, Exatecan mesylate, is a DNA topoisomerase I inhibitor used in cancer research.
  • Deuterium-labeled compound for tracer and internal standard applications.
  • Stable heavy isotopes can affect pharmacokinetic and metabolic profiles.
  • Used in cancer research as a DNA topoisomerase I inhibitor.
  • High purity of 99.8%.
Catalog No. 50-003-52348 Supplier Medchemexpress LLC Supplier No. HY13631AS100MG
Exatecan-d5 mesylate (DX8951f-d5) is a deuterium-labeled compound used as a tracer and internal standard for quantitative analysis by methods like NMR, GC-MS, or LC-MS. Its unlabeled form, Exatecan mesylate, is a DNA topoisomerase I inhibitor used in cancer research.
  • Deuterium-labeled compound for tracer and internal standard applications.
  • Stable heavy isotopes can affect pharmacokinetic and metabolic profiles.
  • Used in cancer research as a DNA topoisomerase I inhibitor.
  • High purity of 99.8%.