Learn More
Medchemexpress LLC Exatecan Intermediate 1 | 110351-94-5 | 100.0% | 263.25 | 1 MG
SDP
Catalog No. 5000404329
Encompass_Preferred
Exatecan Intermediate 1 (compound 6) is an intermediate of Exatecan, a DNA topoisomerase I inhibitor with anticancer effects. Exatecan is a camptothecin analog that acts as the cytotoxic component of ADCs, disrupting tumor cell proliferation and division by interacting with DNA, thereby inhibiting tumor growth. It is primarily used for research of various cancers including ovarian, lung, and breast cancers. This product is for research use only.
  • Intermediate of Exatecan
  • DNA topoisomerase I inhibitor
  • Anticancer effects
  • Camptothecin analog
  • Cytotoxic component of ADCs
  • Disrupts tumor cell proliferation and division
  • Inhibits tumor growth
  • Used for research of various cancers including ovarian, lung, and breast cancers
  • For research use only
Catalog No. 50-004-04329 Supplier Medchemexpress LLC Supplier No. HY42487S1MG
Exatecan Intermediate 1 (compound 6) is an intermediate of Exatecan, a DNA topoisomerase I inhibitor with anticancer effects. Exatecan is a camptothecin analog that acts as the cytotoxic component of ADCs, disrupting tumor cell proliferation and division by interacting with DNA, thereby inhibiting tumor growth. It is primarily used for research of various cancers including ovarian, lung, and breast cancers. This product is for research use only.
  • Intermediate of Exatecan
  • DNA topoisomerase I inhibitor
  • Anticancer effects
  • Camptothecin analog
  • Cytotoxic component of ADCs
  • Disrupts tumor cell proliferation and division
  • Inhibits tumor growth
  • Used for research of various cancers including ovarian, lung, and breast cancers
  • For research use only