Learn More
Medchemexpress LLC Exatecan Intermediate 1 | 110351-94-5 | 100% | 263.25 | 250 MG
SDP
Catalog No. 5000402697
Encompass_Preferred
Exatecan Intermediate 1 (compound 6) is a chemical intermediate used in the synthesis of Exatecan (HY-13631), which is a DNA topoisomerase I inhibitor known for its anticancer properties. It belongs to the class of camptothecin analogs and is primarily used in research to study various cancers such as ovarian, lung, and breast cancers.
  • Serves as the cytotoxic component in Antibody-Drug Conjugates (ADCs).
  • Disrupts tumor cell proliferation and division by interacting with DNA.
  • Suitable for research related to different types of cancers.
Catalog No. 50-004-02697 Supplier Medchemexpress LLC Supplier No. 250MGHY42487
Exatecan Intermediate 1 (compound 6) is a chemical intermediate used in the synthesis of Exatecan (HY-13631), which is a DNA topoisomerase I inhibitor known for its anticancer properties. It belongs to the class of camptothecin analogs and is primarily used in research to study various cancers such as ovarian, lung, and breast cancers.
  • Serves as the cytotoxic component in Antibody-Drug Conjugates (ADCs).
  • Disrupts tumor cell proliferation and division by interacting with DNA.
  • Suitable for research related to different types of cancers.