Learn More
Medchemexpress LLC Exatecan intermediate 13 | 127452-30-6 | 98% | C19H26N2O6 | 250 MG
SDP
Catalog No. 5000358660
Encompass_Preferred
Exatecan intermediate 13 is an analytical standard of Ethyl 2-(6-(acetamidomethyl)-5-oxo-2,3-dihydro-5H-spiro[indolizine-1,2'-dioxolan]-7-yl)butanoate. This product is intended for research and analytical applications, including qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. It is an impurity of Exatecan.
  • Used as an analytical standard (reference standard supplied assay)
  • Suitable for HPLC, GC, and MS applications
  • Intended for research and analytical applications
  • Impurity of Exatecan
Catalog No. 50-003-58660 Supplier Medchemexpress LLC Supplier No. 250MGHY45169
Exatecan intermediate 13 is an analytical standard of Ethyl 2-(6-(acetamidomethyl)-5-oxo-2,3-dihydro-5H-spiro[indolizine-1,2'-dioxolan]-7-yl)butanoate. This product is intended for research and analytical applications, including qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. It is an impurity of Exatecan.
  • Used as an analytical standard (reference standard supplied assay)
  • Suitable for HPLC, GC, and MS applications
  • Intended for research and analytical applications
  • Impurity of Exatecan