Learn More
Medchemexpress LLC Exatecan Intermediate 6 | 143655-58-7 | 98.8% | 235.26 | 100 G
SDP
Catalog No. 5000404179
Encompass_Preferred
Exatecan Intermediate 6 is an intermediate of Exatecan, which is a DNA topoisomerase I inhibitor with an IC50 value of 2.2 μM (0.975 μg/mL) used in cancer research. It can also be used to synthesize Antibody-Drug Conjugates (ADCs).
  • DNA topoisomerase I inhibitor
  • Can be used in cancer research
  • Can synthesize antibody-drug conjugates (ADCs)
  • For research use only
Catalog No. 50-004-04179 Supplier Medchemexpress LLC Supplier No. HY44369100G
Exatecan Intermediate 6 is an intermediate of Exatecan, which is a DNA topoisomerase I inhibitor with an IC50 value of 2.2 μM (0.975 μg/mL) used in cancer research. It can also be used to synthesize Antibody-Drug Conjugates (ADCs).
  • DNA topoisomerase I inhibitor
  • Can be used in cancer research
  • Can synthesize antibody-drug conjugates (ADCs)
  • For research use only