Learn More
Medchemexpress LLC Exatecan Intermediate 6 | 143655-58-7 | 98.98% | 235.26 | 250 MG
SDP
Catalog No. 5000403187
Encompass_Preferred
Exatecan Intermediate 6 is an intermediate of Exatecan and Exatecan. It functions as a DNA topoisomerase I inhibitor, making it suitable for applications in cancer research. This compound can also be utilized for synthesizing Antibody-Drug Conjugates (ADCs).
  • Functions as a DNA topoisomerase I inhibitor
  • Used in cancer research
  • Can be used to synthesize antibody-drug conjugates (ADCs)
Catalog No. 50-004-03187 Supplier Medchemexpress LLC Supplier No. 250MGHY44369
Add to Cart
Add to Cart
Exatecan Intermediate 6 is an intermediate of Exatecan and Exatecan. It functions as a DNA topoisomerase I inhibitor, making it suitable for applications in cancer research. This compound can also be utilized for synthesizing Antibody-Drug Conjugates (ADCs).
  • Functions as a DNA topoisomerase I inhibitor
  • Used in cancer research
  • Can be used to synthesize antibody-drug conjugates (ADCs)