Learn More
Medchemexpress LLC HY-13631A 100mg Medchemexpress, Exatecan (Mesylate) CAS:169869-90-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13631A100MG
Encompass_Preferred
Medchemexpress, HY-13631A 100mg Exatecan (Mesylate) CAS:169869-90-3 Exatecan Mesylate is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL), and can be used in cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9013