Learn More
Medchemexpress LLC Exatecan (mesylate) | 169869-90-3 | 101.1% | 531.55 | 1 ML
SDP
Catalog No. 5000349830
Encompass_Preferred
Exatecan mesylate (DX8951f) is a potent DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). It is a light yellow to green yellow solid that can be used in cancer research. This product is provided as a 10 mM solution in DMSO, ready for laboratory applications.
  • Inhibits DNA topoisomerase I activity
  • Potent activity with an IC50 of 0.975 μg/mL
  • Demonstrates significant inhibition of proliferation in various cancer cell lines, including breast, colon, stomach, and lung cancer
  • Exhibits antitumor activity in in vivo mouse models
  • Stable under recommended storage conditions: 4°C for sealed solid, or -80°C / -20°C for solutions
  • Shipped at room temperature in continental US
Catalog No. 50-003-49830 Supplier Medchemexpress LLC Supplier No. HY13631A10MM/1ML
Exatecan mesylate (DX8951f) is a potent DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). It is a light yellow to green yellow solid that can be used in cancer research. This product is provided as a 10 mM solution in DMSO, ready for laboratory applications.
  • Inhibits DNA topoisomerase I activity
  • Potent activity with an IC50 of 0.975 μg/mL
  • Demonstrates significant inhibition of proliferation in various cancer cell lines, including breast, colon, stomach, and lung cancer
  • Exhibits antitumor activity in in vivo mouse models
  • Stable under recommended storage conditions: 4°C for sealed solid, or -80°C / -20°C for solutions
  • Shipped at room temperature in continental US