Learn More
Medchemexpress LLC Exatecan mesylate | 169869-90-3 | 101.1% | 531.55 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13631A
Encompass_Preferred
Exatecan mesylate (DX8951f) is a DNA topoisomerase I inhibitor primarily used in cancer research. It demonstrates significant inhibition of various cancer cell lines and exhibits antitumor activity in vivo, making it suitable for laboratory centrifugation and storage.
  • DNA topoisomerase I inhibitor
  • Inhibits proliferation of breast, colon, stomach, and lung cancer cells
  • Displays cytotoxic activities against PC-6 and PC-6/SN2-5 cells
  • Exhibits antitumor activity in tumor-bearing mice models
  • Suppresses lymphatic metastasis and eliminates lung metastasis
  • Soluble in H2O at 10 mg/mL and DMSO at 7.41 mg/mL
  • For research use only
Catalog No. 50-003-47936