Learn More
Medchemexpress LLC Exatecan mesylate | 169869-90-3 | MFCD04113012 | >98.0% | C25H26FN3O7S | 10MG
SDP
Catalog No. 5000259214
Encompass_Preferred
Exatecan mesylate is the mesylate salt of exatecan, a potent DNA topoisomerase I inhibitor used in oncology research and as an antibody-drug conjugate payload. It induces DNA damage by stabilizing topoisomerase I-DNA cleavable complexes and has reported in vitro activity (IC50 ≈ 2.2 μM).
Catalog No. 50-002-59214 Supplier Medchemexpress LLC Supplier No. HY13631AG10MG
Exatecan mesylate is the mesylate salt of exatecan, a potent DNA topoisomerase I inhibitor used in oncology research and as an antibody-drug conjugate payload. It induces DNA damage by stabilizing topoisomerase I-DNA cleavable complexes and has reported in vitro activity (IC50 ≈ 2.2 μM).