Learn More
Medchemexpress LLC HY-13631A 500mg , Exatecan (mesylate) CAS:169869-90-3 Purity:>98%
SDP
Catalog No. 502260789
Encompass_Preferred
HY-13631A 500mg Exatecan (mesylate) CAS: 169869-90-3 Exatecan mesylate (DX8951f) is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). Exatecan mesylate can be used in cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0789 Supplier Medchemexpress LLC Supplier No. HY13631A500MG
HY-13631A 500mg Exatecan (mesylate) CAS: 169869-90-3 Exatecan mesylate (DX8951f) is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). Exatecan mesylate can be used in cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.