Learn More
Selleck Chemical LLC Exatecan Mesylate-5mg
SDP
Catalog No. 502389894
Encompass_Preferred
Exatecan Mesylate (DX-8951f) is a water-soluble and non-pro-drug analog of camptothecin and shows significant topoisomerase I inhibition with IC50 of 0.975 μg/ml.
Catalog No. 50-238-9894 Supplier Selleck Chemical LLC Supplier No. S89995MG
Exatecan Mesylate (DX-8951f) is a water-soluble and non-pro-drug analog of camptothecin and shows significant topoisomerase I inhibition with IC50 of 0.975 μg/ml.