Learn More
Medchemexpress LLC HY-13631A 10mg , Exatecan (Mesylate) DX8951f CAS:169869-90-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13631A10MG
Encompass_Preferred
Medchemexpress, HY-13631A 10mg Exatecan (Mesylate) DX8951f CAS:169869-90-3 Formula:C25H26FN3O7S IC50: 2.2 μM (topoisomerase I) Purity:98% Exatecan Mesylate is a water soluble topoisomerase I inhibitor, with an IC 50 of 2.2 μM (0.975 μg/mL), and can be used in cancer research. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4040