Learn More
InVitria Exbumin™ - Recombinant Human Serum Albumin - 100g
Supplier:  InVitria 777HSA097100G
Encompass
InVitria’s Exbumin™ recombinant human serum albumin (rHSA) is a completely animal- and blood-free excipient developed to improve viral stability for vaccines and gene therapies. Exbumin maintains tissue viability during cryopreservation or storage and stabilizes cells and tissue used in cell therapy and regenerative medicine applications. It is an FDA and EMA approved excipient currently used in injectable therapeutics.InVitria manufactures three animal- and blood-free rHSA components: Cellastim S, Exbumin™, and Optibumin. Cellastim S is optimized for cell culture applications. Exbumin is a regulatory approved excipient in final formulations. Optibumin is the highest-purity rHSA available on the market. Made in the USA.
Catalog No. NC1852058
Add to Cart
Request a Quote