Learn More
Med Vet International Zoetis Exced For Swine, 100mg/mL, 100mL
Catalog No. 502828517
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
100mg/ml, 100ml Zoetis Excede for swine (Ceftiofur Crystalline Free Acid). Is a sterile suspension that treats and controls swine respiratory disease for 7 days.
Catalog No. 50-282-8517 Supplier Med Vet International Supplier No. RXEXCEDE
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
100mg/ml, 100ml Zoetis Excede for swine (Ceftiofur Crystalline Free Acid). Is a sterile suspension that treats and controls swine respiratory disease for 7 days.