Learn More
MSC Bulwark Coveralls, Cotton and Nylon, 9.1 cal/cm2, 52 Regular, 1 ea
Catalog No. 5000684567
Encompass
These flame-resistant coveralls provide arc flash protection with an ATPV rating of 9.1 cal./cm2. Designed for comfort and durability, they are constructed from a 6 oz. flame-resistant sateen fabric blend of cotton and nylon.
  • Features a two-way, concealed brass break-away zipper
  • Includes a lay-flat collar and a deep pleated action back for improved mobility
  • Equipped with eight pockets for storage
  • Features elastic waist inserts for an adjustable fit
  • Includes side vent openings with snap closures
  • Designed with concealed snaps at the neck, zipper top, and cuffs
  • Fits chest sizes 50-1/2 in. to 52 in.
  • Meets category 2 arc flash protection requirements
Catalog No. 50-006-84567 Supplier MSC Supplier No. 07931462
These flame-resistant coveralls provide arc flash protection with an ATPV rating of 9.1 cal./cm2. Designed for comfort and durability, they are constructed from a 6 oz. flame-resistant sateen fabric blend of cotton and nylon.
  • Features a two-way, concealed brass break-away zipper
  • Includes a lay-flat collar and a deep pleated action back for improved mobility
  • Equipped with eight pockets for storage
  • Features elastic waist inserts for an adjustable fit
  • Includes side vent openings with snap closures
  • Designed with concealed snaps at the neck, zipper top, and cuffs
  • Fits chest sizes 50-1/2 in. to 52 in.
  • Meets category 2 arc flash protection requirements