Learn More
Sigma Aldrich Fine Chemicals Biosciences EX-CELL(R) 293 Serum-Free Medium for HEK 293 Cells Animal-protein free, with L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture | MFCD01634638 | 1L
Catalog No. NC1255879
Encompass
EX-CELL(R) 293 Serum-Free Medium for HEK 293 Cells Animal-protein free, with L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture | MFCD01634638 | 1L
Catalog No. NC1255879 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 24571C1L
EX-CELL(R) 293 Serum-Free Medium for HEK 293 Cells Animal-protein free, with L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture | MFCD01634638 | 1L