Learn More
Stellar Scientific EXCELBAND 100 BP 3K DNA 500UL
SDP
Catalog No. NC2391338
Encompass_Preferred
The DM2300 ExcelBand 100 bp+3K DNA Ladder is a ready-to-use DNA ladder, which is pre-mixed with loading dye for direct loading on gel. The DM2300 DNA ladder is composed of 12 individual DNA fragments: 3k, 1.5k, 1k, 900, 800, 700, 600, 500, 400, 300, 200 and 100 bp derived from a mixture of PCR products and specifically digested plasmid DNA. This product contains two enhanced bands (1.5 kb and 500 bp) for easy referencing. In addition, two tracking dyes, Xylene cyanol FF and Orange G which mimic the migration of 4,000 bp and 50 bp dsDNA during electrophoresis are also added for real time monitoring.
Catalog No. NC2391338 Supplier Stellar Scientific Supplier No. SMOBDM2300
The DM2300 ExcelBand 100 bp+3K DNA Ladder is a ready-to-use DNA ladder, which is pre-mixed with loading dye for direct loading on gel. The DM2300 DNA ladder is composed of 12 individual DNA fragments: 3k, 1.5k, 1k, 900, 800, 700, 600, 500, 400, 300, 200 and 100 bp derived from a mixture of PCR products and specifically digested plasmid DNA. This product contains two enhanced bands (1.5 kb and 500 bp) for easy referencing. In addition, two tracking dyes, Xylene cyanol FF and Orange G which mimic the migration of 4,000 bp and 50 bp dsDNA during electrophoresis are also added for real time monitoring.