Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE Excelis 4K Camera with attachable 13.3 in. HD Monitor
SDP

Catalog No. 01189360 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-189-360 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-360 Supplier ACCU-SCOPE Supplier No. AU8004KS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delivers high-resolution imaging and documentation with onboard control for streamlined laboratory workflows.

  • High performance 4K delivers full UHD images with exceptional color and resolution
  • CaptaVision+ imaging software adds advanced acquisition image processing measurement report generation plus more
  • The AU8004KS version includes a 13.3 in. 4K monitor that can be mounted directly to the camera allowing easy use of the cameras built-in software with heads-up viewing and space-saving design
  • Intuitive and customizable user interface
  • Simple calibration and measurements
  • Advanced control of exposure gain color balance etc
  • Powerful image processing tools
  • Intelligent flat field correction
  • Record video with voice
  • JPG TIFF PNG DICOM image types
  • Flexible report generation and data export
  • Voice recording with video

Specifications

Type Camera
Autoclavable Not Autoclavable
Color Black
Sterility Non-sterile
Voltage 12 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.