Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ Excelis™ HD Camera with HD Monitor
GREENER_CHOICESDP

Catalog No. 01189214 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Voltage:
12 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01-189-214 12 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-214 Supplier ACCU-SCOPE™ Supplier No. AU600HDS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Excelis™ HD camera delivers true, real-time 1080P HD images at 60 fps, captures 6MP photos and records live video in full HD. The AU-600-HDS version includes a 11.6 in. HD monitor that can be mounted directly to the camera, for easy viewing and a space saving design.

  • True HD imaging for scientific and industrial applications. Capture 6 MP still images.
  • Use the included 11.6 in. HD monitor to view and capture images without a PC.
  • Built-in mouse control software for HDMI viewing and recording without a computer.
  • Simultaneous live image output to an HD monitor and to a PC/Mac via USB2.0 (PC/Mac not included).
  • Perform measurements on live HD images.
  • Fluorescence imaging with high signal-to-noise, exposure times from 1 ms to 10 seconds and 20 gain levels.
  • Monitor mounts directly to the Excelis™ HD camera body for convenient access and doesn't take up valuable bench space.
  • CaptaVision Imaging Software (included) extends the capabilities of the Excelis™ HD with advanced image processing and measurement features (requires PC, not included).NEW Voice Recording with Videos (requires microphone - not included)
  • Standard c-mount easily adapts to most microscopes, 0.35x camera adapter recommended.
  • 1-year warranty.
  • In the box: Excelis™ HD Camera, 11.6 in. HD display, display bracket and screws, HDMI Cable, HDMI adapter, USB 2.0 Cable, Y power splitter cable, 12 V Power Adapter, 8 GB SD Card, USB Mouse, CaptaVision PC Imaging Software.

Specifications

Type Microscope Camera
Autoclavable Not Autoclavable
Sterility Non-sterile
Voltage 12 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.