Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ Excelis™ HD Lite Camera with Monitor
SDP

Catalog No. 01189220 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Voltage:
12 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01-189-220 12 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-220 Supplier ACCU-SCOPE™ Supplier No. AU300HDS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Excelis™ 1080P HD Camera with integrated 11.6 in. HD monitor is a high-fidelity color microscopy camera that delivers true, real-time 1080P HD images for a wide variety of scientific, education and industrial applications. On-board software allows operation without a PC or use with CaptaVision+.

  • HD live imaging for scientific and industrial applications. Capture 5 MP still images.
  • Use the included 11.6 in. HD monitor to view and capture images without a PC.
  • View and capture images without a PC (HDMI monitor required).
  • Built-in mouse control software for HDMI viewing and recording without a computer.
  • Monitor mounts directly to the Excelis™ HD camera body for convenient access and doesn't take up valuable bench space.
  • CaptaVision Imaging Software (included) extends the capabilities of the Excelis™ HD with advanced image processing and measurement features (requires PC, not included).
  • Standard c-mount easily adapts to most microscopes; 0.5x camera adapter recommended.
  • 1-year warranty
  • In the box: Excelis™ HD Camera, 11.6 in. HD display, display bracket and screws, HDMI Cable, HDMI adapter, USB 2.0 Cable, Y power splitter cable, 12 V Power Adapter, 4 GB SD Card, USB Mouse, CaptaVision PC Imaging Software

Specifications

Type Microscope Camera
Autoclavable Not Autoclavable
Sterility Non-sterile
Voltage 12 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.